Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl heptadecanoate-d33, ≥97.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012790 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2790 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2790 Supplier MilliporeSigma Supelco Supplier No. 0088910MG
Add to Cart
Add to Cart

Specifications

CAS 1219804-81-5
Linear Formula C18D33H3O2
MDL Number MFCD03428281
Synonym Heptadecanoic acid-d33 methyl ester
Type Analytical Standard
Quantity 10 mg
Formula Weight 317.68
Percent Purity ≥97.5% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.