Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl heptadecanoate, Thermo Scientific™

Catalog No. AC469300050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469300050 5 g
AC469300010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469300050 Supplier Thermo Scientific Chemicals Supplier No. 469300050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Specifications

Alpha Vector METHYLHEPTADECANOATE
Chemical Name or Material Methyl heptadecanoate
CAS 1731-92-6
Melting Point 29°C to 30°C
Color Colorless or White to Yellow
Boiling Point 152°C to 153°C
Flash Point >110°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C18 H36 O2
MDL Number 00009001
Quantity 5 g
Synonym Methyl margarate
Molecular Weight (g/mol) 284.48
Physical Form Liquid or Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.