Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Heptafluoroisobutyrate 98.0+%, TCI America™
SDP

Catalog No. M20225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20225G 5 g
M202225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20225G Supplier TCI America Supplier No. M20225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 680-05-7
Molecular Formula C5H3F7O2
Molecular Weight (g/mol) 228.066
MDL Number MFCD00633403
InChI Key CGMUKBZUGMXXEF-UHFFFAOYSA-N
Synonym Heptafluoroisobutyric Acid Methyl Ester
PubChem CID 2775518
IUPAC Name methyl 2,3,3,3-tetrafluoro-2-(trifluoromethyl)propanoate
SMILES COC(=O)C(C(F)(F)F)(C(F)(F)F)F

Specifications

Color Colorless
Boiling Point 77°C
Quantity 5 g
UN Number 3272
Formula Weight 228.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Heptafluoroisobutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.