Learn More
Medchemexpress LLC Methyl heptanoate | 106-73-0 | 99.4% | C8H16O2 | 1 KG
SDP
Catalog No. 5000353177
Encompass_Preferred
Methyl heptanoate is a biochemical reagent and organic compound used as a flavoring and fragrance ingredient in the food and cosmetic industries. It also serves as a solvent for various substances, including resins, oils, and waxes, and has been studied for its potential in biofuel and polymer production.
  • Used as a flavoring and fragrance ingredient
  • Acts as a solvent for resins, oils, and waxes
  • Potential for biofuel production
  • Potential for polymer production
  • Biochemical reagent for life science research
Catalog No. 50-003-53177 Supplier Medchemexpress LLC Supplier No. HY1416101KG
Methyl heptanoate is a biochemical reagent and organic compound used as a flavoring and fragrance ingredient in the food and cosmetic industries. It also serves as a solvent for various substances, including resins, oils, and waxes, and has been studied for its potential in biofuel and polymer production.
  • Used as a flavoring and fragrance ingredient
  • Acts as a solvent for resins, oils, and waxes
  • Potential for biofuel production
  • Potential for polymer production
  • Biochemical reagent for life science research