Learn More
Medchemexpress LLC Methyl heptanoate | 106-73-0 | 99.4% | C8H16O2 | 500 G
SDP
Catalog No. 5000357066
Encompass_Preferred
Methyl heptanoate is commonly used as a flavoring and fragrance ingredient in the food and cosmetic industries. It is also utilized as a solvent for various substances, including resins, oils, and waxes. Additionally, it has been studied for its potential as a biofuel and for polymer production applications. Methyl heptanoate is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.
  • Flavoring and fragrance ingredient in food and cosmetic industries
  • Solvent for resins, oils, and waxes
  • Potential for biofuel and polymer production application
  • Biochemical reagent for life science research
Catalog No. 50-003-57066 Supplier Medchemexpress LLC Supplier No. HY141610500G
Methyl heptanoate is commonly used as a flavoring and fragrance ingredient in the food and cosmetic industries. It is also utilized as a solvent for various substances, including resins, oils, and waxes. Additionally, it has been studied for its potential as a biofuel and for polymer production applications. Methyl heptanoate is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.
  • Flavoring and fragrance ingredient in food and cosmetic industries
  • Solvent for resins, oils, and waxes
  • Potential for biofuel and polymer production application
  • Biochemical reagent for life science research