Learn More
Medchemexpress LLC Methyl heptanoate | 106-73-0 | 99.44% | C8H16O2 | 100 G
SDP
Catalog No. 5000356748
Encompass_Preferred
Methyl heptanoate, also known as Methyl enanthate, is a biochemical reagent primarily used as a flavoring and fragrance ingredient in the food and cosmetic industries. It also functions as a solvent for various substances like resins, oils, and waxes, and is being investigated for potential applications in biofuel and polymer production.
  • Used as a flavoring and fragrance ingredient
  • Functions as a solvent for various substances
  • Investigated for biofuel and polymer production applications
  • Serves as a biochemical reagent for life science research
  • Chemically stable under recommended storage conditions
Catalog No. 50-003-56748 Supplier Medchemexpress LLC Supplier No. HY141610100G
Methyl heptanoate, also known as Methyl enanthate, is a biochemical reagent primarily used as a flavoring and fragrance ingredient in the food and cosmetic industries. It also functions as a solvent for various substances like resins, oils, and waxes, and is being investigated for potential applications in biofuel and polymer production.
  • Used as a flavoring and fragrance ingredient
  • Functions as a solvent for various substances
  • Investigated for biofuel and polymer production applications
  • Serves as a biochemical reagent for life science research
  • Chemically stable under recommended storage conditions