Learn More
Medchemexpress LLC Methyl heptanoate | 106-73-0 | MFCD00009537 | 99.44% | C8H16O2 | 25 G
SDP
Catalog No. 5000352543
Encompass_Preferred
Methyl heptanoate, also known as Methyl enanthate, is a biochemical reagent. It is commonly used as a flavoring and fragrance ingredient in the food and cosmetic industries due to its fruity scent. Beyond its use in consumer products, it serves as an effective solvent for a variety of substances, including resins, oils, and waxes. Research also suggests its potential applications in the fields of biofuel and polymer production.
  • Functions as a flavoring and fragrance component.
  • Acts as a versatile solvent for various organic materials.
  • Identified as a biochemical reagent.
  • Has potential for use in biofuel development.
  • Shows promise in polymer synthesis applications.
Catalog No. 50-003-52543 Supplier Medchemexpress LLC Supplier No. 25GHY141610
Methyl heptanoate, also known as Methyl enanthate, is a biochemical reagent. It is commonly used as a flavoring and fragrance ingredient in the food and cosmetic industries due to its fruity scent. Beyond its use in consumer products, it serves as an effective solvent for a variety of substances, including resins, oils, and waxes. Research also suggests its potential applications in the fields of biofuel and polymer production.
  • Functions as a flavoring and fragrance component.
  • Acts as a versatile solvent for various organic materials.
  • Identified as a biochemical reagent.
  • Has potential for use in biofuel development.
  • Shows promise in polymer synthesis applications.