Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl heptanoate | 106-73-0 | MFCD00009537 | 99.44% | C8H16O2 | 50 G
SDP

Catalog No. 5000354867
Encompass_Preferred

Methyl heptanoate is a biochemical reagent commonly used as a flavoring and fragrance ingredient in the food and cosmetic industries. It also serves as a solvent for various substances, including resins, oils, and waxes. Additionally, it has been studied for its potential in biofuel and polymer production applications and is suitable for life science-related research.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Density: 0.87 g/cm³
  • Molecular weight: 144.21
  • Used as a flavoring and fragrance ingredient
  • Utilized as a solvent for various substances
  • Studied for potential in biofuel and polymer production applications
  • Suitable as a biochemical reagent for life science-related research

Catalog No. 50-003-54867 Supplier Medchemexpress LLC Supplier No. HY14161050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl heptanoate is a biochemical reagent commonly used as a flavoring and fragrance ingredient in the food and cosmetic industries. It also serves as a solvent for various substances, including resins, oils, and waxes. Additionally, it has been studied for its potential in biofuel and polymer production applications and is suitable for life science-related research.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Density: 0.87 g/cm³
  • Molecular weight: 144.21
  • Used as a flavoring and fragrance ingredient
  • Utilized as a solvent for various substances
  • Studied for potential in biofuel and polymer production applications
  • Suitable as a biochemical reagent for life science-related research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.