Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl heptanoate, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018696 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8696 5 mL
11-102-0221 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8696 Supplier MilliporeSigma Supelco Supplier No. 752185ML
Add to Cart
Add to Cart

Methyl heptanoate is a fatty acid ester prepared by treating heptanoic acid with methyl alcohol in the presence of hydrochloric acid or sulfuric acid. It is commonly used as a flavoring agent in food products.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 106-73-0
Density 0.880 g/mL (at 20°C)
Flash Point 52°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula C8H16O2
MDL Number MFCD00009537
Boiling Point 171°C to 172°C
Synonym Heptanoic acid methyl ester; Methyl enanthate
Type Analytical Standard
Quantity 5 mL
Formula Weight 144.21
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.