Learn More
Medchemexpress LLC Methyl-Hesperidin | 11013-97-1 | 98.3% | 624.59 | 100 MG
SDP
Catalog No. 5000427880
Encompass_Preferred
Methyl-Hesperidin is a vasodilating agent intended for research use only. It is not for sale to patients. This product is provided in solid form.
  • Vasodilating agent
  • White to light yellow solid appearance
  • For research use only
  • Ships at room temperature
  • Store at 4°C, protected from light
Catalog No. 50-004-27880 Supplier Medchemexpress LLC Supplier No. HYN0165100MG
Methyl-Hesperidin is a vasodilating agent intended for research use only. It is not for sale to patients. This product is provided in solid form.
  • Vasodilating agent
  • White to light yellow solid appearance
  • For research use only
  • Ships at room temperature
  • Store at 4°C, protected from light