Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl-hesperidin | 11013-97-1 | 98.4% | 624.59 g/mol | C29H36O15 | 10 MG
SDP

Catalog No. 5000296632
Encompass_Preferred

Methyl-hesperidin is a flavonoid derivative used as an analytical standard and research reagent; it exhibits vasodilating activity and is provided for laboratory use.

  • Purity: 98.39%.
  • Cas number: 11013-97-1.
  • Molecular weight: 624.59 g/mol.
  • Molecular formula: C29H36O15.
  • Supplied as a dry powder for analytical and in vitro applications.
  • Available in small pack sizes including 10 mg.

Catalog No. 50-002-96632 Supplier Medchemexpress LLC Supplier No. HYN016510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl-hesperidin is a flavonoid derivative used as an analytical standard and research reagent; it exhibits vasodilating activity and is provided for laboratory use.

  • Purity: 98.39%.
  • Cas number: 11013-97-1.
  • Molecular weight: 624.59 g/mol.
  • Molecular formula: C29H36O15.
  • Supplied as a dry powder for analytical and in vitro applications.
  • Available in small pack sizes including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.