Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl-Hesperidin | 11013-97-1 | MFCD01741310 | 98.3% | 624.59 | 50 MG
SDP

Catalog No. 5000427599
Encompass_Preferred

Methyl-Hesperidin is a vasodilating agent. It is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Flavonoids, flavonones, phenols, polyphenols
  • Initial source: Plants, other families
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Purity: 98.29%
  • Molecular weight: 624.59
  • CAS number: 11013-97-1

Catalog No. 50-004-27599 Supplier Medchemexpress LLC Supplier No. HYN016550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl-Hesperidin is a vasodilating agent. It is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Flavonoids, flavonones, phenols, polyphenols
  • Initial source: Plants, other families
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Purity: 98.29%
  • Molecular weight: 624.59
  • CAS number: 11013-97-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.