Learn More
Selleck Chemical LLC Methyl-Hesperidin S2323-50mg
SDP
Catalog No. 503112748
Encompass_Preferred
Methyl Hesperidin is a flavanone glycoside (flavonoid) (C28H34O15) found abundantly in citrus fruits Its aglycone form is called hesperetin
Catalog No. 50-311-2748 Supplier Selleck Chemical LLC Supplier No. S232350MG
Methyl Hesperidin is a flavanone glycoside (flavonoid) (C28H34O15) found abundantly in citrus fruits Its aglycone form is called hesperetin