Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl hexanoate, 99%

Catalog No. AC449240500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449240500 50 g
AC449242500 250 g
AC449240010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC449240500 Supplier Thermo Scientific Chemicals Supplier No. 449240500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 106-70-7 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-70-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009510
InChI Key NUKZAGXMHTUAFE-UHFFFAOYSA-N
Synonym methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
PubChem CID 7824
ChEBI CHEBI:77322
IUPAC Name methyl hexanoate
SMILES CCCCCC(=O)OC

Specifications

Melting Point -71.0°C
Density 0.8840g/mL
Boiling Point 151.0°C
Flash Point 42°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4047 to 1.4067
Linear Formula CH3(CH2)4COOCH3
Quantity 50 g
Specific Gravity 0.884
Solubility Information Solubility in water: insoluble
Viscosity <30 mPa.s (20°C)
Formula Weight 130.18
Percent Purity 99%
Chemical Name or Material Methyl hexanoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-425-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.