Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl homoveratrate | 15964-79-1 | MFCD00017206 | 98.3% | 210.23 g/mol | C11H14O4 | 1 ML
SDP

Catalog No. 5000209255
Encompass_Preferred

Methyl homoveratrate is a small-molecule metabolite of RWJ-26240 that acts as a voltage-sensitive calcium channel blocker. It is supplied as a ready-to-use 10 mM solution in DMSO or as a solid for formulation, analytical, and pharmacological studies.

  • Used for analytical identification in plasma, urine, and fecal extracts.
  • Available as 10 mM solution in DMSO and in multiple solid pack sizes.
  • Purity approximately 98.3% by assay.
  • Molecular formula C11H14O4 and molecular weight 210.23 g/mol.
  • CAS number 15964-79-1 for unambiguous identification.
  • Store pure material frozen; store solutions at low temperature for short term.

Catalog No. 50-002-09255 Supplier Medchemexpress LLC Supplier No. HYW04203910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl homoveratrate is a small-molecule metabolite of RWJ-26240 that acts as a voltage-sensitive calcium channel blocker. It is supplied as a ready-to-use 10 mM solution in DMSO or as a solid for formulation, analytical, and pharmacological studies.

  • Used for analytical identification in plasma, urine, and fecal extracts.
  • Available as 10 mM solution in DMSO and in multiple solid pack sizes.
  • Purity approximately 98.3% by assay.
  • Molecular formula C11H14O4 and molecular weight 210.23 g/mol.
  • CAS number 15964-79-1 for unambiguous identification.
  • Store pure material frozen; store solutions at low temperature for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.