Learn More
Medchemexpress LLC Methyl homoveratrate | 15964-79-1 | MFCD00017206 | 98.3% | 210.23 g/mol | C11H14O4 | 25 G
SDP
Catalog No. 5000208585
Encompass_Preferred
Methyl homoveratrate is an in vivo metabolite of RWJ-26240 (McN5691) used as a reference standard for bioanalytical and pharmacokinetic studies. The compound has formula C11H14O4 and a molecular weight of 210.23 g/mol. It is supplied with documentation for purity and safe handling.
  • High purity for analytical applications.
  • Available in multiple pack sizes, including 25 g.
  • Supplied with certificate of analysis and safety data sheet.
  • Suitable as a metabolite reference in plasma, urine, and fecal analysis.
Catalog No. 50-002-08585 Supplier Medchemexpress LLC Supplier No. 25GHYW042039
Methyl homoveratrate is an in vivo metabolite of RWJ-26240 (McN5691) used as a reference standard for bioanalytical and pharmacokinetic studies. The compound has formula C11H14O4 and a molecular weight of 210.23 g/mol. It is supplied with documentation for purity and safe handling.
  • High purity for analytical applications.
  • Available in multiple pack sizes, including 25 g.
  • Supplied with certificate of analysis and safety data sheet.
  • Suitable as a metabolite reference in plasma, urine, and fecal analysis.