Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl indole-3-carboxylate, 99%, Thermo Scientific™

Catalog No. AC309282500
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309282500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC309282500 Supplier Thermo Scientific Chemicals Supplier No. 309282500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942-24-5
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
InChI Key QXAUTQFAWKKNLM-UHFFFAOYSA-N
Synonym methyl indole-3-carboxylate, indole-3-carboxylic acid methyl ester, methyl 3-indolecarboxylate, 1h-indole-3-carboxylic acid methyl ester, 1h-indole-3-carboxylic acid, methyl ester, 3-carbomethoxyindole, 3-methoxycarbonylindole, methylindole-3-carboxylate, indole-3-carboxylic acid, methyl ester, methyl-3-indolcarboxylate
PubChem CID 589098
ChEBI CHEBI:65019
IUPAC Name methyl 1H-indole-3-carboxylate
SMILES COC(=O)C1=CNC2=CC=CC=C21

Specifications

CAS Min % 96.5
CAS Max % 100.0
Melting Point 149°C to 152°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 g
Formula Weight 175.19
Percent Purity ≥98.5%
Physical Form Amorphous Powder
Chemical Name or Material Methyl indole-3-carboxylate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.