Learn More
Accela Chembio Inc Methyl Indole-4-carboxylate | 25g | 39830-66-5 | MFCD00191222 | 98% | Shelf Life: 1260 Days | Light Sensitive/n2/+4
Catalog No. 50249922
Encompass_Preferred
Methyl Indole-4-carboxylate | 25g | 39830-66-5 | MFCD00191222 | 98% | Shelf Life: 1260 Days | Light Sensitive/n2/+4
Catalog No. 50-249-922 Supplier Accela Chembio Inc Supplier No. SY00275025G
Methyl Indole-4-carboxylate | 25g | 39830-66-5 | MFCD00191222 | 98% | Shelf Life: 1260 Days | Light Sensitive/n2/+4