Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Isobutyl Ketone, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023053 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3053 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-3053 Supplier MilliporeSigma Supelco Supplier No. RTC00009520ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 108-10-1
Melting Point -80°C (literature)
Density 0.801 g/mL (at 25°C (literature))
Boiling Point 117°C to 118°C
Flash Point 14°C (Not Applicable)
Molecular Formula C6H12O
Refractive Index n20/D 1.395 (literature)
Linear Formula (CH3)2 CHCH2COCH3
Quantity 20 mL
MDL Number MFCD00008938
Synonym 4-Methyl-2-pentanone; Isobutyl methyl ketone; Isopropylacetone; MIBK; Methyl isobutyl ketone
Molecular Weight (g/mol) 100.16
Formula Weight 100.16
Grade Certified Reference Material
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.