Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl isobutyrate

Catalog No. 148008500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-800-8500ML 500 mL
14-800-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-800-8500ML Supplier Sigma Aldrich Supplier No. 148008500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Radiofrequency-induced plasmas of methyl isobutyrate has been studied using mass spectrometry.Methyl isobutyrate was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).

Specifications

CAS 547-63-7
Density 0.891 g/mL (at 25°C (literature))
Boiling Point 90°C (lit.)
Molecular Formula C5H10O2
Refractive Index n20/D 1.384 (literature)
Linear Formula (CH3)2 CHCOOCH3
Quantity 500 mL
MDL Number MFCD00008914
Molecular Weight (g/mol) 102.13
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.