Learn More
Selleck Chemical LLC Methyl isoeugenol S3006-100mg
SDP
Catalog No. 503108884
Encompass_Preferred
Methyl isoeugenol (Methylisoeugenol) is an eugenol analogue isoeugenol isolated from Acori Tatarinowii Rhizoma Methyl isoeugenol is a highly attractive lure for Z diversus and would be a valuable inclusion as an attractant in monitoring and male annihilation programs
Catalog No. 50-310-8884 Supplier Selleck Chemical LLC Supplier No. S3006100MG
Methyl isoeugenol (Methylisoeugenol) is an eugenol analogue isoeugenol isolated from Acori Tatarinowii Rhizoma Methyl isoeugenol is a highly attractive lure for Z diversus and would be a valuable inclusion as an attractant in monitoring and male annihilation programs