Learn More
Medchemexpress LLC Methyl isoeugenol | 93-16-3 | 1 ML
SDP
Catalog No. 5000421483
Encompass_Preferred
Methyl isoeugenol (MIE) is a natural food flavor isolated from the leaf of Pimenta pseudocaryophyllus. It exhibits anxiolytic and antidepressant-like effects and is orally active. This product is intended for research use only.
  • Natural food flavor
  • Exhibits anxiolytic and antidepressant-like effects
  • Orally active
  • Cytotoxicity against human KB cells: > 10 μg/mL (IC50)
  • Cytotoxicity against multidrug-resistant human KB-VCR cells: > 10 μg/mL (IC50)
Catalog No. 50-004-21483 Supplier Medchemexpress LLC Supplier No. HYN243910MM/1ML
Methyl isoeugenol (MIE) is a natural food flavor isolated from the leaf of Pimenta pseudocaryophyllus. It exhibits anxiolytic and antidepressant-like effects and is orally active. This product is intended for research use only.
  • Natural food flavor
  • Exhibits anxiolytic and antidepressant-like effects
  • Orally active
  • Cytotoxicity against human KB cells: > 10 μg/mL (IC50)
  • Cytotoxicity against multidrug-resistant human KB-VCR cells: > 10 μg/mL (IC50)