Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl isovanillate | 6702-50-7 | 182.18 | 1 ML
SDP

Catalog No. 5000446688
Encompass_Preferred

Methyl isovanillate is a secondary metabolite isolated from Vitex agnus-castus. It does not exhibit significant activity in the DPPH free radical scavenging experiment.

  • Purity: 99.65%
  • Molecular weight: 182.18
  • Formula: C9H10O4
  • Appearance: Solid
  • CAS number: 6702-50-7
  • Structure classification: Phenols, Monophenols

Catalog No. 50-004-46688 Supplier Medchemexpress LLC Supplier No. HYW00277310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl isovanillate is a secondary metabolite isolated from Vitex agnus-castus. It does not exhibit significant activity in the DPPH free radical scavenging experiment.

  • Purity: 99.65%
  • Molecular weight: 182.18
  • Formula: C9H10O4
  • Appearance: Solid
  • CAS number: 6702-50-7
  • Structure classification: Phenols, Monophenols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.