Learn More
Medchemexpress LLC Methyl isovanillate | 6702-50-7 | 182.18 | 50 G
SDP
Catalog No. 5000448602
Encompass_Preferred
Methyl isovanillate is a secondary metabolite isolated from Vitex agnus-castus. This compound does not exhibit significant activity in the DPPH free radical scavenging experiment and is intended for research use only.
  • Isolated from Vitex agnus-castus
  • Research use only
  • Phenol structure classification
  • Solid appearance
Catalog No. 50-004-48602 Supplier Medchemexpress LLC Supplier No. HYW00277350G
Methyl isovanillate is a secondary metabolite isolated from Vitex agnus-castus. This compound does not exhibit significant activity in the DPPH free radical scavenging experiment and is intended for research use only.
  • Isolated from Vitex agnus-castus
  • Research use only
  • Phenol structure classification
  • Solid appearance