Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl jasmonate

Catalog No. 3927075ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-270-75ML 5 mL
39-270-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-270-75ML Supplier Sigma Aldrich Supplier No. 3927075ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl jasmonate (mj, meja), a methyl ester of jasmonic acid (ja) is a plant growth regulator. it is a fragrant volatile compound isolated from the flowers of jasminum grandiflorum. it has been reported to play an important role in the plant's response to pathogens and wound.methyl jasmonate (mj, meja) is suitable in controlling blue mold decay in sweet cherry fruit caused by penicillium expansum.methyl jasmonate may be used in the following studies:
  • As a potential anti-cancer agent that shows selective cytotoxic effect towards cancer cells
  • To induce the defensive proteinase inhibitor proteins synthesis in plant leaves
  • As an elicitor to enrich the total anthocyanin content (tac) in radish sprouts
  • As an elicitor in inducing the biosynthesis of trans-resveratrol, a plant phenol in vitis vinifera cv. negramaro cell cultures
  • As a mediator of extensive plant transcriptome reprogramming/ remodeling on exogenous treatment to salvia sclarea leaves
  • As a modulator in the expression of chalcone synthase (chs) and proline-rich cell wall protein (prp), two wound-responsive genes in soybean suspension cultures
  • As a starting material in the synthesis of [13c,2h3]-meja, an internal standard used in the quantitative determination of meja in plant tissues
  • As a test compound in the application of reduced graphene oxide-poly(safranine t) film on glassy carbon electrode (rgo-pst/gce) in the electrochemical determination of meja in jasmine essential oil

Specifications

CAS 39924-52-2
Density 1.03 g/mL (at 25°C (literature))
Boiling Point 110°C (0.2 mmHg)
Molecular Formula C13H20O3
Refractive Index n20/D 1.474 (literature)
Linear Formula (O=)C5H6(CH2CH=CHC2H5)CH2CO2CH3
MDL Number MFCD00036656
Quantity 5 mL
Synonym 3-Oxo-2-(2-pentenyl)cyclopentaneacetic acid, methyl ester; Methyl 3-oxo-2-(2-pentenyl)cyclopentaneacetate
Molecular Weight (g/mol) 224.3
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.