Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-Methyl L-aspartate hydrochloride | 16856-13-6 | 98.0% | 183.59 | 5 G
SDP

Catalog No. 5000365972
Encompass_Preferred

β-Methyl L-aspartate hydrochloride is a chemical compound with the formula C5H10ClNO4. It is an L-Aspartic acid derivative, specifically the 4-methyl ester hydrochloride. This product appears as a white to off-white solid and is stable under recommended storage conditions, making it suitable for various laboratory applications.

  • Chemical formula: C5H10ClNO4
  • Molecular weight: 183.59
  • Purity (NMR): 98.0%
  • Appearance: White to off-white solid
  • Melting/freezing point: 191-193°C
  • Stable under recommended storage conditions
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Storage conditions: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Catalog No. 50-003-65972 Supplier Medchemexpress LLC Supplier No. 5GHYY1801
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Methyl L-aspartate hydrochloride is a chemical compound with the formula C5H10ClNO4. It is an L-Aspartic acid derivative, specifically the 4-methyl ester hydrochloride. This product appears as a white to off-white solid and is stable under recommended storage conditions, making it suitable for various laboratory applications.

  • Chemical formula: C5H10ClNO4
  • Molecular weight: 183.59
  • Purity (NMR): 98.0%
  • Appearance: White to off-white solid
  • Melting/freezing point: 191-193°C
  • Stable under recommended storage conditions
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Storage conditions: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.