Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Methyl L-Lactate

Catalog No. 230340100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-034-0100G 100 g
23-034-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 23-034-0100G Supplier Sigma Aldrich Supplier No. 230340100G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Methyl L-lactate can be used as a chiral auxiliary during the synthesis of Bao Gong Teng A, an antiglaucoma compound.

Specifications

CAS 27871-49-4
Density 1.09 g/mL at 25°C
Boiling Point 144°C to 145°C
Molecular Formula C4H8O3
Refractive Index n20/D 1.413
Linear Formula CH3CH(OH)CO2CH3
MDL Number MFCD00064265
Quantity 100 g
Synonym Methyl (S)-(-)-lactate
Molecular Weight (g/mol) 104.1 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.