Learn More
Medchemexpress LLC Methyl L-pyroglutamate | 4931-66-2 | 99.6% | 143.14 | 5 G
SDP
Catalog No. 5000401322
Encompass_Preferred
Methyl L-pyroglutamate, also known as (S)-Methyl 5-oxopyrrolidine-2-carboxylate or L-Pyroglutamic acid methyl ester, is a compound isolated from P. oleracea. It exhibits anti-inflammatory activity and is intended for research use only.
  • Isolated from P. oleracea.
  • Exhibits anti-inflammatory activity.
  • Intended for research use only.
  • Purity of 99.6%.
Catalog No. 50-004-01322 Supplier Medchemexpress LLC Supplier No. HY322915G
Methyl L-pyroglutamate, also known as (S)-Methyl 5-oxopyrrolidine-2-carboxylate or L-Pyroglutamic acid methyl ester, is a compound isolated from P. oleracea. It exhibits anti-inflammatory activity and is intended for research use only.
  • Isolated from P. oleracea.
  • Exhibits anti-inflammatory activity.
  • Intended for research use only.
  • Purity of 99.6%.