Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Lactate, +98%, Spectrum™ Chemical
SDP

Catalog No. 18621064 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
50 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-064 500 mL
18-621-062 25 mL
18-621-063 100 mL
18-621-065 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-621-064 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1716500ML
Add to Cart
Add to Cart

Synonym: Methyl 2-Hydroxypropanoate, Methyl DL-Lactate, DL-Methyl Lactate

Chemical Identifiers

CAS 547-64-8

Specifications

Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.