Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Lactate, +98%, Spectrum™ Chemical
SDP

Catalog No. 18621065 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
50 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-065 50 L
18-621-062 25 mL
18-621-063 100 mL
18-621-064 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-621-065 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M171650LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

Synonym: Methyl 2-Hydroxypropanoate, Methyl DL-Lactate, DL-Methyl Lactate

Chemical Identifiers

CAS 547-64-8

Specifications

Packaging Drum
Quantity 50 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.