Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl laurate

Catalog No. 502899065 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-9065 25 g
23-459-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-289-9065 Supplier Sigma Aldrich Supplier No. 23459125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl laurate
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-82-0
Melting Point 4°C to 5°C (lit.)
Density 0.87 g/mL (at 25°C (literature))
Boiling Point 262°C (766 mmHg)
Molecular Formula C13H26O2
Refractive Index n20/D 1.431 (literature)
Linear Formula CH3(CH2)10 CO2CH3
MDL Number MFCD00008966
Quantity 25 g
Synonym Lauric acid methyl ester; Methyl dodecanoate
Molecular Weight (g/mol) 214.34
Percent Purity 99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.