Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl laurate | 111-82-0 | 214.34 | 1 KG
SDP

Catalog No. 5000448739
Encompass_Preferred

Methyl laurate is a 12-carbon saturated fatty acid, which is an esterified version of lauric acid. It is intended for research use only.

  • Appearance as a colorless to light yellow liquid
  • Purity of 99.9%
  • Molecular weight of 214.34
  • Synonyms include methyl dodecanoate
  • Chemical formula is C13H26O2

Catalog No. 50-004-48739 Supplier Medchemexpress LLC Supplier No. HYW0042861KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl laurate is a 12-carbon saturated fatty acid, which is an esterified version of lauric acid. It is intended for research use only.

  • Appearance as a colorless to light yellow liquid
  • Purity of 99.9%
  • Molecular weight of 214.34
  • Synonyms include methyl dodecanoate
  • Chemical formula is C13H26O2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.