Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl laurate | 111-82-0 | 214.34 | 500 G
SDP

Catalog No. 5000445460
Encompass_Preferred

Methyl laurate, also known as methyl dodecanoate, is an esterified 12-carbon saturated fatty acid. It appears as a colorless to light yellow liquid with a density of 0.87 g/cm³. This chemical is intended for laboratory research and substance manufacturing.

  • Esterified 12-carbon saturated fatty acid
  • Colorless to light yellow liquid appearance
  • Density of 0.87 g/cm³
  • Melting/freezing point: 4-5°C
  • Boiling point: 263°C at 760 mmHg
  • Flash point: 110°C
  • Stable under recommended storage conditions
  • For research use only

Catalog No. 50-004-45460 Supplier Medchemexpress LLC Supplier No. HYW004286500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl laurate, also known as methyl dodecanoate, is an esterified 12-carbon saturated fatty acid. It appears as a colorless to light yellow liquid with a density of 0.87 g/cm³. This chemical is intended for laboratory research and substance manufacturing.

  • Esterified 12-carbon saturated fatty acid
  • Colorless to light yellow liquid appearance
  • Density of 0.87 g/cm³
  • Melting/freezing point: 4-5°C
  • Boiling point: 263°C at 760 mmHg
  • Flash point: 110°C
  • Stable under recommended storage conditions
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.