Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl laurate, 98+%

Catalog No. p-3758436
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
250 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215560025 2.5 g Glass bottle
AC215560100 10 g Glass bottle
AC215560500 50 g Glass bottle
AC215562500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC215560025 Supplier Thermo Scientific Chemicals Supplier No. 215560025
Only null left
Add to Cart

CAS: 111-82-0 | C13H26O2 | 214.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.35
MDL Number MFCD00008966
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Melting Point 4°C to 5°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 261°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330
Linear Formula CH3(CH2)10CO2CH3
Quantity 2.5 g
Beilstein 02,361
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble
Formula Weight 214.35
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl laurate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 203-911-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.