Learn More
Medchemexpress LLC Methyl levulinate | 624-45-3 | 25 G
SDP
Catalog No. 5000420716
Encompass_Preferred
Methyl levulinate is an Alkaloids product isolated from the stems and leaves of *Ervatamia hainanensis*. It is commonly used as a laboratory chemical and in the manufacture of various substances.
  • Isolated from the stems and leaves of *Ervatamia hainanensis*
  • Appears as a colorless to light yellow liquid
  • Has a purity of 99.94%
  • Features a molecular weight of 130.14
  • Chemical formula is C6H10O3
Catalog No. 50-004-20716 Supplier Medchemexpress LLC Supplier No. 25GHYN3272
Methyl levulinate is an Alkaloids product isolated from the stems and leaves of *Ervatamia hainanensis*. It is commonly used as a laboratory chemical and in the manufacture of various substances.
  • Isolated from the stems and leaves of *Ervatamia hainanensis*
  • Appears as a colorless to light yellow liquid
  • Has a purity of 99.94%
  • Features a molecular weight of 130.14
  • Chemical formula is C6H10O3