Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl levulinate | 624-45-3 | 500 G
SDP

Catalog No. 5000417911
Encompass_Preferred

Methyl levulinate (NSC 24874) is a liquid laboratory chemical identified as an Alkaloids product. It can be isolated from the stems and leaves of Ervatamia hainanensis and is used in the manufacture of substances. It has a relative density of 1.02 g/cm³ and is for research use only.

  • Purity of 99.94%
  • Appears as a colorless to light yellow liquid
  • Molecular weight: 130.14
  • Chemical formula: C6H10O3
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition

Catalog No. 50-004-17911 Supplier Medchemexpress LLC Supplier No. HYN3272500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl levulinate (NSC 24874) is a liquid laboratory chemical identified as an Alkaloids product. It can be isolated from the stems and leaves of Ervatamia hainanensis and is used in the manufacture of substances. It has a relative density of 1.02 g/cm³ and is for research use only.

  • Purity of 99.94%
  • Appears as a colorless to light yellow liquid
  • Molecular weight: 130.14
  • Chemical formula: C6H10O3
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.