Learn More
Medchemexpress LLC Methyl linoleate | 112-63-0 | 294.47 | 1 G
SDP
Catalog No. 5000416536
Encompass_Preferred
Methyl linoleate, a major active constituent of Sageretia thea fruit (HFSF), is a significant anti-melanogenic compound. It functions by downregulating microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins.
  • Downregulates microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins.
  • Reduces melanogenesis through the Akt/GSK3β signaling pathway.
  • Inhibits β-catenin-mediated transcriptional activation of MITF.
Catalog No. 50-004-16536 Supplier Medchemexpress LLC Supplier No. HYN14811G
Methyl linoleate, a major active constituent of Sageretia thea fruit (HFSF), is a significant anti-melanogenic compound. It functions by downregulating microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins.
  • Downregulates microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins.
  • Reduces melanogenesis through the Akt/GSK3β signaling pathway.
  • Inhibits β-catenin-mediated transcriptional activation of MITF.