Learn More
Medchemexpress LLC Methyl linoleate | 112-63-0 | 294.47 | 5 G
SDP
Catalog No. 5000416513
Encompass_Preferred
Methyl linoleate is a major active constituent of Sageretia thea fruit (HFSF) and has been identified as a significant anti-melanogenic compound. It functions by downregulating microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins.
  • Suitable for research use only
  • Downregulates melanogenesis via the Akt/GSK3β signaling pathway
  • Inhibits β-catenin-mediated transcriptional activation of MITF
Catalog No. 50-004-16513 Supplier Medchemexpress LLC Supplier No. HYN14815G
Methyl linoleate is a major active constituent of Sageretia thea fruit (HFSF) and has been identified as a significant anti-melanogenic compound. It functions by downregulating microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins.
  • Suitable for research use only
  • Downregulates melanogenesis via the Akt/GSK3β signaling pathway
  • Inhibits β-catenin-mediated transcriptional activation of MITF