Learn More
Selleck Chemical LLC Methyl linoleate
SDP
Catalog No. 5000003368
Encompass_Preferred
Methyl linoleate (Linoleic acid methyl ester) found in cloves is a fatty acid methyl ester of linoleic acid which is an essential fatty acid in mammalian nutrition and is used in the biosynthesis of prostaglandins and cell membranes
Catalog No. 50-000-03368 Supplier Selleck Chemical LLC Supplier No. S575925UL
Methyl linoleate (Linoleic acid methyl ester) found in cloves is a fatty acid methyl ester of linoleic acid which is an essential fatty acid in mammalian nutrition and is used in the biosynthesis of prostaglandins and cell membranes