Learn More
Selleck Chemical LLC Methyl linolenate
SDP
Catalog No. 5000003361
Encompass_Preferred
Methyl linolenate (Linolenic acid methyl ester Methyl alpha-linolenate) is a fatty acid methyl ester derived from alpha-linolenic acid and acts as a plant metabolite and an insect attractant
Catalog No. 50-000-03361 Supplier Selleck Chemical LLC Supplier No. S56825MG
Methyl linolenate (Linolenic acid methyl ester Methyl alpha-linolenate) is a fatty acid methyl ester derived from alpha-linolenic acid and acts as a plant metabolite and an insect attractant