Learn More
Medchemexpress LLC Methyl linolenate | 301-00-8 | 99.5% | 292.46 | 1 G
SDP
Catalog No. 5000400686
Encompass_Preferred
Methyl linolenate is a polyunsaturated fatty acid (PUFA). It exhibits anti-melanogenesis activity with an IC50 of 60 μM and can be utilized for studies on the mechanisms and prevention of oxidation/peroxidation of unsaturated fatty acids.
  • Polyunsaturated fatty acid (PUFA).
  • Anti-melanogenesis activity with an IC50 of 60 μM.
  • Useful for studies of the mechanisms and prevention of oxidation/peroxidation of unsaturated fatty acids.
Catalog No. 50-004-00686 Supplier Medchemexpress LLC Supplier No. HY212681G
Methyl linolenate is a polyunsaturated fatty acid (PUFA). It exhibits anti-melanogenesis activity with an IC50 of 60 μM and can be utilized for studies on the mechanisms and prevention of oxidation/peroxidation of unsaturated fatty acids.
  • Polyunsaturated fatty acid (PUFA).
  • Anti-melanogenesis activity with an IC50 of 60 μM.
  • Useful for studies of the mechanisms and prevention of oxidation/peroxidation of unsaturated fatty acids.