Learn More
Methyl -linolenate Solution, ≥98.5% (GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 16326-32-2 |
| Flash Point | 13°C - Closed Cup |
| Linear Formula | C19H32O2 |
| MDL Number | MFCD00010455 |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 292.46 |
| Percent Purity | ≥98.5% (GC) |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 250 mg/mL in ethanol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.