Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl malonyl chloride

Catalog No. 6340610ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-406-10ML 10 mL
63-406-50ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-406-10ML Supplier Sigma Aldrich Supplier No. 6340610ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl malonyl chloride can be used in the synthesis of methyl 7-hydroxy[1,3]thiazolo[5,4-b]pyridin-5(4h)-one-6-carboxylates and 6-carboxamides, which can act like hiv integrase strand transfer inhibitors
  • It can also be used in the synthesis of strychnos alkaloids like (-)-leuconicine a and (-)-leuconicine b

Specifications

CAS 37517-81-0
Density 1.273 g/mL (at 25°C (literature))
Boiling Point 57°C to 59°C (12 mmHg)
Molecular Formula C4H5ClO3
Refractive Index n20/D 1.432 (literature); n20/D 1.432
Linear Formula CH3OCOCH2COCl
MDL Number MFCD00013657
Quantity 10 mL
Synonym Methyl chloroformylacetate
Molecular Weight (g/mol) 136.53
Percent Purity ≥97.0% (AT)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.