Learn More
Grainger TCI America Methyl Methacrylate Polymer, White, Crystal-Powder, 25 g
Catalog No. 502780918
Encompass_Preferred
Methyl Methacrylate Polymer, also known as PMMA or poly(methyl methacrylate), is a transparent thermoplastic utilized for various research applications. It serves as a versatile material, commonly employed as a lightweight or shatter-resistant alternative to traditional glass.
  • Solid crystal powder form
  • Autoignition temperature of 304°C
  • Soluble in toluene and tetrahydrofuran (THF)
  • Identified by CAS RN 9011-14-7
  • Non-hazardous for transportation under DOT, IATA, and IMDG regulations
  • Included on the EPA Toxic Substances Control Act (TSCA) inventory
Catalog No. 50-278-0918 Supplier Grainger Supplier No. 19KT11
Methyl Methacrylate Polymer, also known as PMMA or poly(methyl methacrylate), is a transparent thermoplastic utilized for various research applications. It serves as a versatile material, commonly employed as a lightweight or shatter-resistant alternative to traditional glass.
  • Solid crystal powder form
  • Autoignition temperature of 304°C
  • Soluble in toluene and tetrahydrofuran (THF)
  • Identified by CAS RN 9011-14-7
  • Non-hazardous for transportation under DOT, IATA, and IMDG regulations
  • Included on the EPA Toxic Substances Control Act (TSCA) inventory