Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI America Methyl Methacrylate Polymer, White Powder, 500 g

Catalog No. 502781496
Encompass_Preferred

Methyl methacrylate polymer is a high-molecular-weight solid reagent suitable for laboratory research applications. This synthetic material, also known as PMMA, is provided as a white powder or crystal and is soluble in solvents such as toluene and tetrahydrofuran. It is stable under normal conditions and is not classified as a hazardous substance.

  • Physical state: solid powder or crystal
  • Molecular formula: [C5H8O2]n
  • Average molecular weight: approx. 800,000
  • Solubility: soluble in toluene and tetrahydrofuran
  • Storage conditions: recommended in a cool and shaded area, typically below 15°C
  • Regulatory compliance: TSCA listed and DSL compliant

Catalog No. 50-278-1496 Supplier Grainger Supplier No. 19KT12
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl methacrylate polymer is a high-molecular-weight solid reagent suitable for laboratory research applications. This synthetic material, also known as PMMA, is provided as a white powder or crystal and is soluble in solvents such as toluene and tetrahydrofuran. It is stable under normal conditions and is not classified as a hazardous substance.

  • Physical state: solid powder or crystal
  • Molecular formula: [C5H8O2]n
  • Average molecular weight: approx. 800,000
  • Solubility: soluble in toluene and tetrahydrofuran
  • Storage conditions: recommended in a cool and shaded area, typically below 15°C
  • Regulatory compliance: TSCA listed and DSL compliant

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.