Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Methacrylate (Stabilized/Certified ACS), Fisher Chemical

Catalog No. o36294 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O3629-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. O3629-4
This item is not returnable. View return policy

Chemical Identifiers

CAS 80-62-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC

Specifications

Melting Point -48°C
Color Colorless
Density 0.930g/cm³
Boiling Point 100°C
Flash Point 7°C
Assay Percent Range ≥98 %
Packaging Glass Bottle
Linear Formula CH2:C(CH3)COOCH3
Quantity 4 L
UN Number 1247
Stabilizer 20 to 30ppm
Vapor Density 3.5g/mol
Formula Weight 100.12
Percent Purity ≥98%
Grade Certified
Physical Form Liquid
Chemical Name or Material Methyl Methacrylate, Stabilized
Show More Show Less
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.