Learn More
Selleck Chemical LLC Methyl methanesulfonate
SDP
Catalog No. 503137035
Encompass_Preferred
Methyl methanesulfonate (MMS) a DNA alkylating agent modifies both guanine (to 7-methylguanine) and adenine (to 3-methlyladenine) to cause base mispairing and replication blocks respectively
Catalog No. 50-313-7035 Supplier Selleck Chemical LLC Supplier No. E06091G
Methyl methanesulfonate (MMS) a DNA alkylating agent modifies both guanine (to 7-methylguanine) and adenine (to 3-methlyladenine) to cause base mispairing and replication blocks respectively