Learn More
TARGETMOL CHEMICALS INC METHYL METHANESULFONATE 200MG
SDP
Catalog No. 502578914
Encompass_Preferred
Also available in 500 mg 1000 mg and bulk. Please contact Fisher for quotes. Methyl methanesulfonate is a DNA alkylator that modifies adenine (3-methyladenine) and guanine (7-methyladenine) respectively resulting in base mismatch and replication disorders. purity: 98%
Catalog No. 50-257-8914 Supplier TARGETMOL CHEMICALS INC Supplier No. T9985200MG
Also available in 500 mg 1000 mg and bulk. Please contact Fisher for quotes. Methyl methanesulfonate is a DNA alkylator that modifies adenine (3-methyladenine) and guanine (7-methyladenine) respectively resulting in base mismatch and replication disorders. purity: 98%