Learn More
Medchemexpress LLC Methyl methanesulfonate | 66-27-3 | MFCD00007557 | 110.13 | 500 G
SDP
Catalog No. 5000446021
Encompass_Preferred
Methyl methanesulfonate is an alkylating agent that transfers methyl groups and induces DNA damage. It is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Transfers methyl groups and induces DNA damage.
  • Can be used as a biological material or organic compound for life science-related research.
  • High purity.
  • Induces necroptosis in A549 cells and exhibits cytotoxicity against human MCF7 cells.
  • Induces DNA damage in mouse models, increasing DNA with comet tails in liver, lung, kidney, and spleen.
Catalog No. 50-004-46021 Supplier Medchemexpress LLC Supplier No. HYW004702500G
Methyl methanesulfonate is an alkylating agent that transfers methyl groups and induces DNA damage. It is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Transfers methyl groups and induces DNA damage.
  • Can be used as a biological material or organic compound for life science-related research.
  • High purity.
  • Induces necroptosis in A549 cells and exhibits cytotoxicity against human MCF7 cells.
  • Induces DNA damage in mouse models, increasing DNA with comet tails in liver, lung, kidney, and spleen.