Learn More
Medchemexpress LLC Methyl methanesulfonate (Standard) | 66-27-3 | 110.13 | 25 G
SDP
Catalog No. 5000448084
Encompass_Preferred
Methyl methanesulfonate is an alkylating agent that transfers methyl groups and induces DNA damage. It is a biochemical reagent used as a biological material or organic compound for life science-related research.
  • Induces DNA damage
  • Transfers methyl groups as an alkylating agent
  • Biochemical reagent for life science-related research
  • Colorless to light yellow liquid appearance
  • Density of 1.3 g/cm³
Catalog No. 50-004-48084 Supplier Medchemexpress LLC Supplier No. 25GHYW004702
Methyl methanesulfonate is an alkylating agent that transfers methyl groups and induces DNA damage. It is a biochemical reagent used as a biological material or organic compound for life science-related research.
  • Induces DNA damage
  • Transfers methyl groups as an alkylating agent
  • Biochemical reagent for life science-related research
  • Colorless to light yellow liquid appearance
  • Density of 1.3 g/cm³